Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113303.1 | NP_001106774.1 | 1 - 2 | 258 | 97 | |
| GenBank mRNA | BC105140.1 | - | 1 - 2 | 258 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105140.1, NM_001113303.1 transcripts.
Set Membership: |
|

DYNLL2
Genomic Map