Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113303.1 | NP_001106774.1 | 2 - 3 | 393 | 95 | |
| GenBank mRNA | BC105140.1 | - | 2 - 3 | 393 | 95 | |
More Information
Set Membership: |
|

DYNLL2
Genomic Map