Assay Details
| Gene Symbol: NDUFA4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113305.2 | NP_001106776.1 | 3 - 4 | 255 | 73 | |
| GenBank mRNA | BC103157.1 | - | 3 - 4 | 256 | 73 | |
More Information
Set Membership: |
|

NDUFA4L2
Genomic Map