Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113313.2 | NP_001106784.1 | 1 - 2 | 157 | 61 | |
| GenBank mRNA | BC112751.1 | - | 1 - 2 | 158 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112751.1, NM_001113313.2 transcripts.
Set Membership: |
|

NRGN
Genomic Map