Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113313.2 | NP_001106784.1 | 2 - 3 | 384 | 157 | |
| GenBank mRNA | BC112751.1 | - | 2 - 3 | 385 | 157 | |
More Information
Set Membership: |
|

NRGN
Genomic Map