Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113313.2 | NP_001106784.1 | 3 - 4 | 409 | 53 | |
| GenBank mRNA | BC112751.1 | - | 3 - 4 | 410 | 53 | |
More Information
Set Membership: |
|

NRGN
Genomic Map