Assay Details
| Gene Symbol: ATP5MPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113315.1 | NP_001106786.1 | 1 - 2 | 71 | 125 | |
| GenBank mRNA | BC102582.1 | - | 1 - 2 | 71 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102582.1, NM_001113315.1 transcripts.
Set Membership: |
|

ATP5MPL
Genomic Map