Assay Details
| Gene Symbol: ATP5MPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113315.1 | NP_001106786.1 | 3 - 4 | 212 | 111 | |
| GenBank mRNA | BC102582.1 | - | 3 - 4 | 212 | 111 | |
More Information
Set Membership: |
|

ATP5MPL
Genomic Map