Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113316.2 | NP_001106787.1 | 3 - 4 | 311 | 59 | |
| GenBank mRNA | BC102970.1 | - | 3 - 4 | 314 | 59 | |
More Information
Set Membership: |
|

HSBP1
Genomic Map