Assay Details
| Gene Symbol: UQCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113723.2 | NP_001107195.1 | 1 - 2 | 197 | 61 | |
| GenBank mRNA | BC102720.1 | - | 1 - 2 | 198 | 61 | |
More Information
Set Membership: |
|

UQCR10
Genomic Map