Assay Details
| Gene Symbol: ANAPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113724.2 | NP_001107196.1 | 1 - 2 | 513 | 97 | |
| GenBank mRNA | BC102426.1 | - | 1 - 2 | 514 | 97 | |
More Information
Set Membership: |
|

ANAPC11
Genomic Map