Assay Details
| Gene Symbol: BRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114510.2 | NP_001107982.1 | 1 - 2 | 125 | 64 | |
| GenBank mRNA | BC102222.1 | - | 1 - 2 | 126 | 64 | |
More Information
Set Membership: |
|

BRK1
Genomic Map