Assay Details
| Gene Symbol: CAMK2N2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114514.2 | NP_001107986.1 | 1 - 2 | 233 | 95 | |
| GenBank mRNA | BC140670.1 | - | 1 - 2 | 234 | 95 | |
More Information
Set Membership: |
|

CAMK2N2
Genomic Map