Assay Details
| Gene Symbol: SFTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114523.2 | NP_001107995.1 | 1 - 2 | 164 | 79 | |
| GenBank mRNA | BC126804.1 | - | 1 - 2 | 167 | 79 | |
More Information
Set Membership: |
|

SFTA2
Genomic Map