Assay Details
| Gene Symbol: P2RY13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098152.1 | NP_001091621.1 | 1 | 2653 | 118 | |
| GenBank mRNA | BC140571.1 | - | 1 | 2653 | 118 | |
More Information
Set Membership: |
|

P2RY13
Genomic Map