Assay Details
| Gene Symbol: RNASEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098165.1 | NP_001091634.1 | 6 - 7 | 2290 | 97 | |
| GenBank mRNA | BC142502.1 | - | 6 - 7 | 2247 | 97 | |
| DQ497162.1 | - | 6 - 7 | 2290 | 97 | ||
More Information
Set Membership: |
|

RNASEL
Genomic Map