Assay Details
| Gene Symbol: ADSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099192.1 | NP_001092662.1 | 6 - 7 | 597 | 99 | |
| GenBank mRNA | BC142211.1 | - | 6 - 7 | 597 | 99 | |
More Information
Set Membership: |
|

ADSS1
Genomic Map