Assay Details
| Gene Symbol: PNPLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099200.1 | NP_001092670.1 | 6 - 7 | 1842 | 74 | |
| GenBank mRNA | BC142232.1 | - | 6 - 7 | 1842 | 74 | |
More Information
Set Membership: |
|

PNPLA1
Genomic Map