Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099205.1 | NP_001092675.1 | 2 - 3 | 422 | 71 | |
| GenBank mRNA | BC146214.1 | - | 2 - 3 | 422 | 71 | |
More Information
Set Membership: |
|

LARP6
Genomic Map