Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099210.2 | NP_001092680.1 | 3 - 4 | 298 | 98 | |
| GenBank mRNA | BC142374.1 | - | 3 - 4 | 300 | 98 | |
| BC148018.1 | - | 3 - 4 | 282 | 98 | ||
More Information
Set Membership: |
|

RPS17
Genomic Map