Assay Details
| Gene Symbol: LRRC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099395.1 | NP_001092865.1 | 1 - 2 | 152 | 79 | |
| GenBank mRNA | BC146155.1 | - | 1 - 2 | 152 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146155.1, NM_001099395.1 transcripts.
Set Membership: |
|

LRRC3B
Genomic Map