Assay Details
| Gene Symbol: CYSLTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099726.1 | NP_001093196.1 | 1 - 2 | 159 | 101 | |
| GenBank mRNA | BC142477.1 | - | 1 - 2 | 159 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142477.1, NM_001099726.1 transcripts.
Set Membership: |
|

CYSLTR1
Genomic Map