Assay Details
| Gene Symbol: LOC787410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099729.2 | NP_001093199.1 | 7 - 8 | 743 | 102 | |
| GenBank mRNA | BC142433.1 | - | 7 - 8 | 744 | 102 | |
More Information
Set Membership: |
|

LOC787410
Genomic Map