Assay Details
| Gene Symbol: LOC787410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099729.2 | NP_001093199.1 | 11 - 12 | 1224 | 119 | |
| GenBank mRNA | BC142433.1 | - | 11 - 12 | 1225 | 119 | |
More Information
Set Membership: |
|

LOC787410
Genomic Map