Assay Details
| Gene Symbol: TRAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101280.1 | NP_001094750.1 | 1 - 2 | 235 | 89 | |
| GenBank mRNA | BC151614.1 | - | 1 - 2 | 235 | 89 | |
More Information
Set Membership: |
|

TRAF4
Genomic Map