Assay Details
| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101286.1 | NP_001094756.1 | 1 - 2 | 48 | 103 | |
| GenBank mRNA | BC148048.1 | - | 1 - 2 | 48 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148048.1, NM_001101286.1 transcripts.
Set Membership: |
|

CEP20
Genomic Map