Assay Details
| Gene Symbol: GPR65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101303.2 | NP_001094773.1 | 1 - 2 | 80 | 132 | |
| GenBank mRNA | BC151280.1 | - | 1 - 2 | 81 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151280.1, NM_001101303.2 transcripts.
Set Membership: |
|

GPR65
Genomic Map