Assay Details
| Gene Symbol: TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101306.2 | NP_001094776.1 | 1 - 2 | 409 | 92 | |
| GenBank mRNA | BC149528.1 | - | 1 - 2 | 410 | 92 | |
More Information
Set Membership: |
|

TNFRSF6B
Genomic Map