Assay Details
| Gene Symbol: FIZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101308.1 | NP_001094778.1 | 2 - 3 | 449 | 69 | |
| GenBank mRNA | BC149069.1 | - | 2 - 3 | 449 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149069.1, NM_001101308.1 transcripts.
Set Membership: |
|

FIZ1
Genomic Map