Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102327.1 | 1989 | 90 | |||
| GenBank mRNA | BC148939.1 | - | 1989 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10D| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102327.1 | 1989 | 90 | |||
| GenBank mRNA | BC148939.1 | - | 1989 | 90 | ||
Set Membership: |
|