Assay Details
| Gene Symbol: ELOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102333.2 | NP_001095803.2 | 3 - 4 | 381 | 72 | |
| GenBank mRNA | BC149490.1 | - | 3 - 4 | 253 | 72 | |
More Information
Set Membership: |
|

ELOA
Genomic Map