Assay Details
Gene Symbol: DEFB122A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102339.1 | NP_001095809.1 | 1 - 2 | 61 | 65 | |
GenBank mRNA | BC108176.1 | - | 105 | 65 | ||
EU036212.1 | - | 1 - 2 | 61 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
