Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102341.2 | NP_001095811.1 | 5 - 6 | 751 | 105 | |
| GenBank mRNA | BC148136.1 | - | 5 - 6 | 752 | 105 | |
More Information
Set Membership: |
|

LGALS3
Genomic Map