Assay Details
| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102351.1 | NP_001095821.1 | 7 - 8 | 1619 | 111 | |
| GenBank mRNA | BC148938.1 | - | 7 - 8 | 1619 | 111 | |
More Information
Set Membership: |
|

BTNL2
Genomic Map