Assay Details
| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102355.1 | NP_001095825.1 | 6 - 7 | 773 | 73 | |
| GenBank mRNA | BC151529.1 | - | 6 - 7 | 773 | 73 | |
More Information
Set Membership: |
|

AGXT
Genomic Map