Assay Details
| Gene Symbol: RAVER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103108.1 | NP_001096578.1 | 10 - 11 | 1721 | 64 | |
| GenBank mRNA | BC123485.1 | - | 10 - 11 | 1721 | 64 | |
More Information
Set Membership: |
|

RAVER1
Genomic Map