Assay Details
| Gene Symbol: SNX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103328.1 | NP_001096798.1 | 2 - 3 | 385 | 60 | |
| GenBank mRNA | BC151344.1 | - | 2 - 3 | 385 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151344.1, NM_001103328.1 transcripts.
Set Membership: |
|

SNX21
Genomic Map