Assay Details
| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103333.2 | NP_001096803.2 | 6 - 7 | 1461 | 59 | |
| GenBank mRNA | BC134591.1 | - | 6 - 7 | 1463 | 59 | |
More Information
Set Membership: |
|

CCDC157
Genomic Map