Assay Details
| Gene Symbol: B3GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103343.1 | NP_001096813.1 | 12 - 13 | 1746 | 82 | |
| GenBank mRNA | BC151379.1 | - | 12 - 13 | 1746 | 82 | |
More Information
Set Membership: |
|

B3GALNT2
Genomic Map