Assay Details
| Gene Symbol: TEAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103347.1 | NP_001096817.1 | 9 - 10 | 780 | 61 | |
| GenBank mRNA | BC122637.1 | - | 9 - 10 | 780 | 61 | |
More Information
Set Membership: |
|

TEAD2
Genomic Map