Assay Details
| Gene Symbol: IK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103349.1 | NP_001096819.1 | 16 - 17 | 1448 | 69 | |
| GenBank mRNA | BC123466.1 | - | 16 - 17 | 1448 | 69 | |
More Information
Set Membership: |
|

IK
Genomic Map