Assay Details
| Gene Symbol: LYPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105024.1 | NP_001098494.1 | 4 - 5 | 592 | 63 | |
| GenBank mRNA | BC149085.1 | - | 4 - 5 | 592 | 63 | |
More Information
Set Membership: |
|

LYPD5
Genomic Map