Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105025.2 | NP_001098495.1 | 1 - 2 | 237 | 89 | |
| GenBank mRNA | BC149509.1 | - | 1 - 2 | 238 | 89 | |
More Information
Set Membership: |
|

LRR1
Genomic Map