Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105025.2 | NP_001098495.1 | 2 - 3 | 342 | 123 | |
| GenBank mRNA | BC149509.1 | - | 2 - 3 | 343 | 123 | |
More Information
Set Membership: |
|

LRR1
Genomic Map