Assay Details
| Gene Symbol: PCYT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105052.2 | NP_001098522.1 | 7 - 8 | 990 | 72 | |
| GenBank mRNA | BC112538.1 | - | 7 - 8 | 993 | 72 | |
More Information
Set Membership: |
|

PCYT1A
Genomic Map