Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105461.1 | NP_001098931.1 | 1 - 2 | 342 | 137 | |
| GenBank mRNA | BC123837.1 | - | 1 - 2 | 342 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123837.1, NM_001105461.1 transcripts.
Set Membership: |
|

FLACC1
Genomic Map