Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105461.1 | NP_001098931.1 | 5 - 6 | 785 | 63 | |
| GenBank mRNA | BC123837.1 | - | 5 - 6 | 785 | 63 | |
More Information
Set Membership: |
|

FLACC1
Genomic Map