Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105461.1 | NP_001098931.1 | 8 - 9 | 1041 | 73 | |
| GenBank mRNA | BC123837.1 | - | 8 - 9 | 1041 | 73 | |
More Information
Set Membership: |
|

FLACC1
Genomic Map