Assay Details
| Gene Symbol: ATAD3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105462.1 | NP_001098932.1 | 4 - 5 | 470 | 62 | |
| GenBank mRNA | BC134487.1 | - | 4 - 5 | 470 | 62 | |
More Information
Set Membership: |
|

ATAD3A
Genomic Map