Assay Details
| Gene Symbol: GLMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105476.1 | NP_001098946.1 | 1 - 2 | 117 | 78 | |
| GenBank mRNA | BC119876.1 | - | 1 - 2 | 117 | 78 | |
More Information
Set Membership: |
|

GLMP
Genomic Map